missing translation for 'onlineSavingsMsg'
Learn More

Mefenpyr-diethyl 98.0+%, TCI America™

$177.08 - $531.25

Chemical Identifiers

CAS 135590-91-9
Molecular Formula C16H18Cl2N2O4
Molecular Weight (g/mol) 373.23
MDL Number MFCD09753375
InChI Key OPGCOAPTHCZZIW-UHFFFAOYSA-N
Synonym Diethyl 1-(2,4-Dichlorophenyl)-4,5-dihydro-5-methyl-1H-pyrazole-3,5-dicarboxylate, 1-(2,4-Dichlorophenyl)-4,5-dihydro-5-methyl-1H-pyrazole-3,5-dicarboxylic Acid Diethyl Ester, Diethyl 1-(3,4-Dichlorophenyl)-5-methyl-2-pyrazoline-3,5-dicarboxylate, 1-(3,4-
PubChem CID 10937610
IUPAC Name diethyl 1-(2,4-dichlorophenyl)-5-methyl-4H-pyrazole-3,5-dicarboxylate
SMILES CCOC(=O)C1=NN(C(C1)(C)C(=O)OCC)C2=C(C=C(C=C2)Cl)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M26041G
View Documents
TCI America
M26041G
1 g
Each for $177.08
Only null left
 
M26045G
View Documents
TCI America
M26045G
5 g
Each for $531.25
Only null left
 
Description

Description

Specifications

Chemical Identifiers

135590-91-9
373.23
OPGCOAPTHCZZIW-UHFFFAOYSA-N
10937610
CCOC(=O)C1=NN(C(C1)(C)C(=O)OCC)C2=C(C=C(C=C2)Cl)Cl
C16H18Cl2N2O4
MFCD09753375
Diethyl 1-(2,4-Dichlorophenyl)-4,5-dihydro-5-methyl-1H-pyrazole-3,5-dicarboxylate, 1-(2,4-Dichlorophenyl)-4,5-dihydro-5-methyl-1H-pyrazole-3,5-dicarboxylic Acid Diethyl Ester, Diethyl 1-(3,4-Dichlorophenyl)-5-methyl-2-pyrazoline-3,5-dicarboxylate, 1-(3,4-
diethyl 1-(2,4-dichlorophenyl)-5-methyl-4H-pyrazole-3,5-dicarboxylate

Specifications

135590-91-9
White-Yellow
MFCD09753375
3077
OPGCOAPTHCZZIW-UHFFFAOYSA-N
diethyl 1-(2,4-dichlorophenyl)-5-methyl-4H-pyrazole-3,5-dicarboxylate
10937610
≥98.0% (HPLC)
Mefenpyr-diethyl
52°C
C16H18Cl2N2O4
1 g
Diethyl 1-(2,4-Dichlorophenyl)-4,5-dihydro-5-methyl-1H-pyrazole-3,5-dicarboxylate, 1-(2,4-Dichlorophenyl)-4,5-dihydro-5-methyl-1H-pyrazole-3,5-dicarboxylic Acid Diethyl Ester, Diethyl 1-(3,4-Dichlorophenyl)-5-methyl-2-pyrazoline-3,5-dicarboxylate, 1-(3,4-
CCOC(=O)C1=NN(C(C1)(C)C(=O)OCC)C2=C(C=C(C=C2)Cl)Cl
373.23
373.23
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : UQ6473000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'