missing translation for 'onlineSavingsMsg'
Learn More

Mefenpyr-diethyl 98.0+%, TCI America™

Catalog No. M26041G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26041G 1 g
M26045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26041G Supplier TCI America Supplier No. M26041G

Chemical Identifiers

CAS 135590-91-9
Molecular Formula C16H18Cl2N2O4
Molecular Weight (g/mol) 373.23
MDL Number MFCD09753375
InChI Key OPGCOAPTHCZZIW-UHFFFAOYSA-N
Synonym Diethyl 1-(2,4-Dichlorophenyl)-4,5-dihydro-5-methyl-1H-pyrazole-3,5-dicarboxylate, 1-(2,4-Dichlorophenyl)-4,5-dihydro-5-methyl-1H-pyrazole-3,5-dicarboxylic Acid Diethyl Ester, Diethyl 1-(3,4-Dichlorophenyl)-5-methyl-2-pyrazoline-3,5-dicarboxylate, 1-(3,4-
PubChem CID 10937610
IUPAC Name diethyl 1-(2,4-dichlorophenyl)-5-methyl-4H-pyrazole-3,5-dicarboxylate
SMILES CCOC(=O)C1=NN(C(C1)(C)C(=O)OCC)C2=C(C=C(C=C2)Cl)Cl

Specifications

Melting Point 52°C
Color White-Yellow
Quantity 1 g
UN Number 3077
Formula Weight 373.23
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Mefenpyr-diethyl
RTECSNumber UQ6473000
TSCA No