Learn More
Medix™ Replacement Capillary Packs for TIC Kits
Catalog No. 23230237
Encompass
Change view
Click to view available options
For Use With (Application):
Leukotic kits
Thrombotic kits
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
23230237 Leukotic kits
23230236 Thrombotic kits
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23230237 Supplier Medix™ Supplier No. BLR4013
Includes
100 pieces of volume capillaries and 100 pieces chamber filling capillaries
Replacement Capillary Packs for leukotic, Blu+ WBC and thrombotic kits
Specifications
For Use With (Equipment) Leukotic kits
Product Type Replacement Capillary Pack
For Use With (Application) Leukotic kits
Includes 100 Pieces of volume capillaries and 100 pieces chamber filling capillaries