missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Melanocyte Basal Medium Kit, calcium free (formerly Medium 254CF Kit)
GREENER_CHOICE

Catalog No. M254CF500
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M254CF500 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M254CF500 Supplier Gibco™ Supplier No. M254CF500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Gibco™ Human Melanocyte Basal Medium Kit (formerly Medium 254CF, Kit) enables the culture of human epidermal melanocytes.

The Gibco Human Melanocyte Basal Medium Kit, Calcium Free (formerly Medium 254CF Kit), enables the culture of human epidermal melanocytes.

The kit contains two components: 
• Human Melanocyte Basal Medium, Calcium Free (formerly Medium 254CF)—a calcium-free version of Human Melanocyte Basal Medium (formerly Medium 254) for those who desire to vary the concentration of calcium 
• A sterile stock solution of calcium chloride (0.2 M, 0.5 mL)

Human Melanocyte Basal Medium, Calcium Free, is a basal medium containing essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts.

This medium does not contain antibiotics, antimycotics, hormones, growth factors, or proteins. This medium is HEPES and bicarbonate buffered and is designed for use in an incubator with an atmosphere of 5% CO2/95% air.

To support plating and long-term proliferation of normal human melanocytes, Human Melanocyte Basal Medium, Calcium Free, must be supplemented with calcium plus either Human Melanocyte Growth Supplement (HMGS, Cat. No. S-002-5) or PMA-Free Human Melanocyte Growth Supplement-2 (HMGS-2, Cat. No. S-016-5). Each of these supplements contains all of the growth factors, hormones, and tissue extracts necessary for growth of melanocytes in Human Melanocyte Basal Medium, Calcium Free.

Note: Calcium chloride must be added to this medium prior to use. Calcium concentration from other sources is 0.5 μM in unsupplemented Human Melanocyte Basal Medium, Calcium Free.

Specifications

Cell Type Melanocytes
Form Liquid
Product Type Medium 254CF Kit
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Phenol Red
Without Additives No Calcium Chloride
Product Line Cascade Biologics
Quantity 1 Kit
Shipping Condition Wet ice
Content And Storage
  • 500 mL Human Melanocyte Basal Medium, Calcium Free (formerly “Medium 254CF”); store in refrigerator (2–8°C) and protect from light
  • Sterile stock solution of calcium chloride (0.2 M, 0.5 mL); store in refrigerator (2–8°C)
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.