missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Large Vessel Endothelial Cell Basal Medium (formerly Medium 200), Phenol Red Free
GREENER_CHOICE

Catalog No. M200PRF500
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M200PRF500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M200PRF500 Supplier Gibco™ Supplier No. M200PRF500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Medium 200PRF

Human Large Vessel Endothelial Cell Basal Medium (formerly 'Medium 200') is a sterile, liquid tissue culture medium intended for use as one component in a complete culture environment for normal human large vessel endothelial cells. When supplemented with low serum growth supplement (LSGS), this medium supports the plating and proliferation of endothelial cells at culture densities between 2.5 x 103 cells/cm2 and 1 x 105 cells/cm2. Additional applications may include primary isolation of endothelial cells. This medium is also available with phenol red (Cat. No. M200500).

Human Large Vessel Endothelial Cell Basal Medium is a basal culture medium containing essential and non-essential amino acids, vitamins, other organic compounds, trace minerals, and inorganic salts. This medium does not contain antibiotics, antimycotics, hormones, growth factors, or proteins. It is HEPES and bicarbonate buffered and designed for use in an incubator with an atmosphere of 5% CO2/95% air. To support the plating and proliferation of human large vessel endothelial cells, this medium must be supplemented with LSGS (Cat. No. S-003-10; kit = Cat. No. S-003-K).

Specifications

Cell Type Endothelial Cells
Form Liquid
Product Type Basal Medium
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives HEPES
Without Additives No Phenol Red
Product Line Cascade Biologics
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage 4 degrees C
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.