Learn More
Corning™ Medium 199 (Mod.)
Catalog No. MT10060CV
Change view
Click to view available options
Quantity:
6 x 500 mL
Catalog No. Quantity
MT10060CV 6 x 500 mL
1 options
Catalog No. MT10060CV Supplier Corning™ Supplier No. 10060CV
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Guaranteed low endotoxin levels ≤0.025 EU/mL
  • Complete line of media and reagents for mammalian in vitro cell culture
  • Ready-to-use media is supplied in polyethylene teraphthalate (PET) plastic bottles
  • Sonically welded cap liners eliminate toxic resins or glues that could leach into media
  • Shipped at ambient temperature
Specifications
Product Type M199/EBSS
Quantity 6 x 500 mL
pH 7 to 7.4