missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Medium 154CF, Kit
GREENER_CHOICE

Catalog No. M154CF500
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M154CF500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M154CF500 Supplier Gibco™ Supplier No. M154CF500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Medium 154CF, Kit

A sterile, liquid medium for the culture of human epidermal keratinocytes. Medium 154CF is Medium 154 prepared without calcium chloride. Calcium chloride is provided as a separate component with each bottle of medium. This basal medium requires the addition of HKGS (S-001-5) or HKGS Kit (S-001-K) prior to use.

Specifications

Cell Type Keratinocyte Cells
Classification Serum-free
Form Liquid
Product Type Medium 154CF Kit
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Phenol Red
Without Additives No Calcium Chloride
Product Line Cascade Biologics
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage 4 degrees C
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.