missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Fibroblast Expansion Basal Medium (formerly Medium 106)
GREENER_CHOICE

Catalog No. M106500
Encompass
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M106500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M106500 Supplier Gibco™ Supplier No. M106500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Human Fibroblast Expansion Basal Medium (formerly Medium 106) is a sterile, liquid tissue culture medium intended for use as one component in a complete culture environment for the growth of normal human dermal fibroblasts.

This basal culture medium contains:

  • Essential and non-essential amino acids
  • Vitamins
  • Other organic compounds
  • Trace minerals
  • Inorganic salts.

Human Fibroblast Expansion Basal Medium does NOT contain:

  • Antibiotics
  • Antimycotics
  • Hormones
  • Growth factors
  • Proteins

This medium is HEPES and bicarbonate buffered and is designed for use in an incubator with an atmosphere of 5% CO2 / 95% air. To support the plating and long-term proliferation of human dermal fibroblasts, Human Fibroblast Expansion Basal Medium must be supplemented with Low Serum Growth Supplement (LSGS, Cat. No. S-003-10), or Low Serum Growth Supplement Kit (LSGS Kit, Cat. No. S-003-K).

Specifications

Cell Type Fibroblast Cells
Form Liquid
Product Type Medium 106
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives HEPES, Phenol Red
Product Line Cascade Biologics
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Store at 4°C.
Culture Type Mammalian Cell Culture
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.