missing translation for 'onlineSavingsMsg'
Learn More

MediPurpose™ Safety Lancet

Catalog No. 22000102
Change view
Click to view available options
Capacity (Metric):
20 to 40μL
40 to 60μL
Color:
Orange
Pink
Includes:
Needle
Needle Gauge:
21g
22g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Color Includes Needle Gauge
22000102 40 to 60μL Pink Needle 21g
22000101 20 to 40μL Orange Needle 22g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22000102 Supplier MediPurpose™ Supplier No. SLN300100015

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Safety needle or blade lancet is safely concealed before and after use

  • Unique, one-step activation mechanism for fingerstick sampling—no arming is required
  • Once lancet is used, it is rendered inoperative, providing added safety for patient and clinician
  • High-speed delivery and penetration method minimizes patient pain and clinician error
  • Latex-free
  • Available with either a needle or blade, each with different depths of penetration, to provide optimal blood flow for different applications and skin types

Specifications

Includes Needle
Product Type Safety Lancet
Capacity (English) 0.00135 to 0.00202 oz.
For Use With (Application) For all POC tests requiring whole Blood samples, including Glucose, cholesterol, HIV and more
Depth (English) 0.11 in.
Certifications/Compliance OSHA
Capacity (Metric) 40 to 60μL
Capacity 0.00135 to 0.00202 oz., 40 to 60μL
Depth (Metric) 2.8mm
Quantity 100 Pack
Needle Gauge 21g
Color Pink
Show More Show Less