missing translation for 'onlineSavingsMsg'
Learn More

MediPurpose™ Safety Lancet

Catalog No. 22000101
Change view
Click to view available options
Capacity (Metric):
20 to 40μL
40 to 60μL
Color:
Orange
Pink
Includes:
Needle
Needle Gauge:
21g
22g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Color Includes Needle Gauge
22000101 20 to 40μL Orange Needle 22g
22000102 40 to 60μL Pink Needle 21g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22000101 Supplier MediPurpose™ Supplier No. SLN240

Safety needle or blade lancet is safely concealed before and after use

  • Unique, one-step activation mechanism for fingerstick sampling—no arming is required
  • Once lancet is used, it is rendered inoperative, providing added safety for patient and clinician
  • High-speed delivery and penetration method minimizes patient pain and clinician error
  • Latex-free
  • Available with either a needle or blade, each with different depths of penetration, to provide optimal blood flow for different applications and skin types

Specifications

Certifications/Compliance OSHA
Capacity 0.00067 to 0.00135 oz., 20 to 40μL
Capacity (Metric) 20 to 40μL
Depth (English) 0.08 in.
For Use With (Application) For all POC tests requiring whole Blood samples, including Glucose, cholesterol, HIV and more
Product Type Safety Lancet
Quantity 100 Pack
Includes Needle
Color Orange
Capacity (English) 0.00067 to 0.00135 oz.
Depth (Metric) 2.2mm
Needle Gauge 22g
Show More Show Less