missing translation for 'onlineSavingsMsg'
Learn More

Medicom™ SafeBasics™ Lab Gowns

Catalog No. p-4940010
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
296359544 Medium
19910554 Small
19910556 Large
19910557 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 296359544 Supplier Medicom™ Supplier No. MED8017/DUP
Only null left

Added protection from cross-contamination.

  • Increased breathability with 3-ply SMS fabric
  • Comfortable fit with knitted cuffs and collar
  • Front snap closure
  • Fluid resistant material
  • 10 per bag, 5 bags per case

Specifications

Product Type General Purpose Lab Coat, Single-use Lab Coat
Size Medium
Length (Metric) 101 cm
Length (English) 39.7 in.
For Use With (Application) For protection from cross contamination
No. of Pockets 2
Gender Unisex
Material SMS
Color Sky Blue
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Latex Free Yes
Autoclavable Not Autoclavable
Show More Show Less