missing translation for 'onlineSavingsMsg'
Learn More

Medegen Sharps Container, Stackable

Stackable Sharps Container with transparent top for visual identification of fill level.

Supplier:  Medegen 8706T

 View more versions of this product

Catalog No. 14-827-18

Only null left
Explore available promotions

Description

Description

  • Needle removal port for easy disposal
  • Locking lids for transport
  • Made of autoclavable polypropylene to meet industry standards for puncture resistance
Specifications

Specifications

3.8 L
Translucent
Single-use
Yes
Yes
25.4 cm
10 in.
6.7 in.
Sharps Container
1 gal.
7 x 10 x 6.7 in.
17 cm
17.8 cm
Non-sterile
Autoclavable
7 in.
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions