missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medegen Biohazardous Waste Bags, LLDPE Film, Coreless Roll
These Biohazardous Waste Bags pass ASTMD1709-85, 165 Gram Dart Test.
$339.48
Specifications
| Color | Red, Black |
|---|---|
| For Use With (Application) | Used to collect, store and transport biohazardous and infectious waste within the health care facility, outpatient surgery center, alternate care clinic, physician's office and lab |
| Includes | Biohazardous or infectious waste with biohazard symbol |
| Material | LLDPE |
| Product Type | Biohazardous Waste Bag |
| Catalog Number | Dimensions (L x W) | Capacity (English) | Thickness (English) | Certifications/Compliance | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Dimensions (L x W) | Capacity (English) | Thickness (English) | Certifications/Compliance | Price | Quantity & Availability | |||
|
1482743B
|
30.5 x 41 in. | 30 gal. | 1.2 mil | ASTM Dart |
Case of 250 Each for $339.48
|
|
|||
Description
- Made from low-density polyethylene
- Minimize the risk of contamination from biohazardous materials during collection and transport
- Bags are red with ''Biohazardous Waste/Infectious Waste'' and biohazard symbol imprinted in black ink
- Individually folded
Specifications
| Red, Black | |
| Biohazardous or infectious waste with biohazard symbol | |
| Biohazardous Waste Bag |
| Used to collect, store and transport biohazardous and infectious waste within the health care facility, outpatient surgery center, alternate care clinic, physician's office and lab | |
| LLDPE |
May NOT be used in CA, FL, PA or for interstate (between state) transport.