missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medegen Biohazardous Waste Bags, Autoclavable
These Biohazardous Waste Bags are autoclavable.
$315.35 - $315.35
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Certifications/Compliance | Meets 165 g Dart Impact Resistance Test |
| Includes | Indicator, flat pack, twist ties |
| Material | Polypropylene |
| Product Type | Autoclavable Biohazardous Waste Bag |
| Catalog Number | Mfr. No. | Dimensions (L x W) | Color | Thickness (Metric) | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Dimensions (L x W) | Color | Thickness (Metric) | Price | Quantity | |||
|
14-828-350
|
N/A | 38 x 47 in. | Clear | 0.045 mm |
Case of 100 Each for $315.35
|
|
|||
Description
- Autoclavable
Specifications
| Autoclavable | |
| Indicator, flat pack, twist ties | |
| Autoclavable Biohazardous Waste Bag |
| Meets 165 g Dart Impact Resistance Test | |
| Polypropylene |