Learn More
Bellco Glass Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spot. On s
Supplier: BeLLCo Glass 563760050
Manufactured from Type 1 Class A Borosilicate Glass. These media storage bottles are chemically resistant ans stable. They are suitable for storage as well as for mixing and sampling. They are autoclavable and have white graduations and marking spot. On s