Learn More
Ohaus Media Bottle Clamp
Description
Designed to hold Erlenmeyer flasks from 10 mL to 6 L. Constructed of type 302 and 304 stainless steel. Includes hardware for easy attachment to platforms. Flask clamps 50 mL and higher are supplied with a spring to hold the flask in place. The 2.8 L clamp is designed to hold a Fernbach flask. Media bottle clamps feature the same details as flask clamps.
Specifications
Specifications
| Product Type | Media Bottle Clamp |
| For Use With (Equipment) | Erlenmeyer Flasks from 10 mL to 6 L |
| Size | 500 mL |