missing translation for 'onlineSavingsMsg'
Learn More

Meclofenamic acid sodium salt, 99+%

Catalog No. p-7041852
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6048403 1 g
AAJ6048406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6048403 Supplier Thermo Scientific Chemicals Supplier No. J6048403
Only null left

Meclofenamic acid (meclofenamate, CAS # 644-62-2) is a nonsteroidal anti-inflammatory drug (NSAID) with antipyretic and antineoplastic activities. | CAS: 6385-02-0 | C14H13Cl2NNaO2 | 321.153 g/mol

Inhibits 5-lipoxygenase and cyclooxygenase; NSAID

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Meclofenamic acid is an aminobenzoic acid-based NSAID that is characterized by two aryl moieties

• Meclofenamic acid is an inhibitor of arachidonate 5-lipoxygenase and prostaglandin-endoperoxide synthase

• Meclofenamic acid prevents pyrogen release and inhibits wound granulation; it may also act as a neoplastic

Application

• Meclofenamic acid can complex metals, making it of interest to drug delivery studies that target its toxic metal complexes to cancerous cell types

• It is suitable for laboratory research on pain, degeneration, wound healing, and tumor growth

Chemical Identifiers

CAS 6385-02-0
Molecular Formula C14H13Cl2NNaO2
Molecular Weight (g/mol) 321.153
MDL Number MFCD00077376
InChI Key JKTYCTJLWZYQGM-UHFFFAOYSA-N
Synonym meclofenamate sodium
PubChem CID 131673979
IUPAC Name 2-(2,6-dichloro-3-methylanilino)benzoic acid;molecular hydrogen;sodium
SMILES [HH].CC1=C(C(=C(C=C1)Cl)NC2=CC=CC=C2C(=O)O)Cl.[Na]

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water at 50mg/ml; Also soluble in DMF, DMSO or ethanol
Formula Weight 318.13
Percent Purity ≥99%
Physical Form Crystalline Solid
Chemical Name or Material Meclofenamic acid sodium salt
EINECSNumber 228-983-3
RTECSNumber CB2975500
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only