missing translation for 'onlineSavingsMsg'
Learn More

Mecamylamine hydrochloride

Catalog No. aaj60257mb
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ60257MB 25 mg
AAJ60257MC 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ60257MB Supplier Thermo Scientific Chemicals Supplier No. J60257MB

Nicotinyl acetylcholine receptor antagonist | CAS: 826-39-1 | C11H22ClN | 203.754 g/mol

Nicotinyl acetylcholine receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 826-39-1
Molecular Formula C11H22ClN
Molecular Weight (g/mol) 203.754
MDL Number MFCD00151462
InChI Key PKVZBNCYEICAQP-CIISUUNXSA-N
Synonym mecamylamine hcl, 2-methylamino isocamphane hydrochloride, n,2,3,3-tetramethylbicyclo 2.2.1 heptan-2-amine hydrochloride, a: 600/1 223m072 melphalan hydrochloride 3223-07-2, 1s,4r-n,2,3,3-tetramethylbicyclo 2.2.1 heptan-2-amine hydrochloride, mecamylamine hydrochloride, united states pharmacopeia usp reference standard
PubChem CID 25162921
IUPAC Name (1R,4S)-N,2,2,3-tetramethylbicyclo[2.2.1]heptan-3-amine;hydrochloride
SMILES CC1(C2CCC(C2)C1(C)NC)C.Cl

Specifications

Color White
Quantity 25 mg
UN Number UN2811
Merck Index 14,5774
Formula Weight 203.75
Physical Form Powder
Chemical Name or Material Mecamylamine hydrochloride
Hazard Category H301-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 212-555-8
RTECSNumber RB6900000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only