Designed to bring simplicity to pH testing of meat products, the HI99163 Meat pH Meter is engineered to withstand the tough environment of meat processing plants. The specialized FC2323 probe features a PVDF body, conical tip with stainless steel blade, and open junction making it ideal for the measurement of pH in meat processing.0.2 pH accuracyComes with all the necessary solutions and batteries- everything you need to get started measuring right away.Specialized electrode for spot checking the pH during meat processing.