missing translation for 'onlineSavingsMsg'
Learn More

United Scientific™ Measuring Cylinder, To Deliver (TD), Class A, Unserialized

Catalog No. S15995
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S15995 5 mL
S15738 10 mL
S16096 25 mL
S15973 50 mL
S15967 100 mL
S16020 250 mL
S16019 500 mL
S16049 1000 mL
S15736 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. S15995 Supplier United Scientific™ Supplier No. UNBCCTD5
Only null left

Cylinders are made from borosilicate glass and ideal for everyday lab use.

  • United Scientific™ graduated cylinders comply with ASTM E1272, Class A standards, batch certified.
  • Each cylinder features a double metric scale
  • Borosilicate glass 3.3 cylinders with pour spouts feature heavy uniform wall tubing and attached glass hexagonal bases.
  • All cylinders include bumper guards and a batch calibration certificate.
  • Strain Point 515°C, Annealing Point 565°C, Softening Point 820°C

Specifications

Tolerance Metric ASTM E1272
Type Graduated Cylinder
Class A
Material Borosilicate Glass (3.3)
Capacity (Metric) 5 mL
Graduations 0.1 mL