missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ Measurement Kit
Description
- Reusable wooden storage box.
- Comprehensive, teacher-developed activity guide features 15 different activities.
Specifications
Specifications
| Grade | 5+ |
| For Use With (Application) | Variety of tools and objects for teaching basics of measurement |
| Includes | Depth gauge, micrometer, plastic protractor, 12 in. plastic ruler, tape measure, brass vernier caliper, wire gauge with conversion table, aluminum square, aluminum wire, graduated cylinder, plastic cylinder, plastic vial |
| Class Size | 1 Lab Group |
| Product Type | Measurement Kit |