Learn More
Selleck Chemical Llc Enzalutamide (MDV3100) 50mg 915087-33-1 DFS Item
Supplier:  Selleck Chemicals Co Ltd S125050MG
Enzalutamide (MDV3100) is an androgen-receptor (AR) antagonist with IC50 of 36 nM in LNCaP cells. Enzalutamide is shown to increase autophagy. *For Research & Development use only. Product is not intended for drug, household, or human consumption.
Catalog No. 507018
Only null left
Explore available promotions