Learn More
SignalChem Biotech Cyclin Y Protein 100ug
Catalog No. 502575435
Encompass
Recombinant human Cyclin Y (2-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 502575435 Supplier SignalChem Biotech Supplier No. C8730G100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant human Cyclin Y (2-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.