missing translation for 'onlineSavingsMsg'
Learn More

MDEPAP 95.0+%, TCI America™

Catalog No. A54001G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A54001G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A54001G Supplier TCI America Supplier No. A54001G
Only null left

[Extraction-spectrophotometric reagent for anionic surfactants]

Chemical Identifiers

CAS 74920-80-2
Molecular Formula C16H21IN4
Molecular Weight (g/mol) 396.276
MDL Number MFCD00082424
InChI Key FBXMWVQMHYGKQY-UHFFFAOYSA-M
Synonym 1-Methyl-4-(4-diethylaminophenylazo)pyridinium Iodide
PubChem CID 14366671
IUPAC Name N,N-diethyl-4-[(1-methylpyridin-1-ium-4-yl)diazenyl]aniline;iodide
SMILES CCN(CC)C1=CC=C(C=C1)N=NC2=CC=[N+](C=C2)C.[I-]

Specifications

Melting Point 190°C
Color Black-Gray
Quantity 1 g
Formula Weight 396.28
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material MDEPAP [=1-Methyl-4-(4-diethylaminophenylazo)pyridinium Iodide] [Extraction-spectrophotometri