missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Velp Scientific MCT-W20-V015-A Block
Description
- Compatible with Velp Scientific ECODryBlock
- Suitable for molecular biology workflows, DNA/RNA extraction, protein assays, and sample incubation
- Tube type: 1.5 mL microcentrifuge
- Number of wells: 20
- Well depth: 36.9 mm
- Well size: Ø 11.5 mm
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 1.5 mL |
| For Use With (Equipment) | Microcentrifuge Tubes |
| Material | Aluminum |
| No. of Places | 20 |
| Type | Dry Block |
| Tube Size | 1.5 mL |
| Dimensions (L x W x H) | 95 x 76 x 51 mm |
| Depth (Metric) | 36.9 mm |
| Diameter (Metric) Well | 11.5 mm |