Learn More
ACROBiosystems Inc Recombinant Protein;Human M-CSF R / CSF1R / CD115 Protein;HEK293;200UG;CSR,CSF1R,C-FMS,CD115,CSFR,FIM2,FMS,M-CSFR,CSF-1R,CSF1 R
Catalog No. 501657528
Encompass
Recombinant Protein;Human M-CSF R / CSF1R / CD115 Protein;200UG;Human M-CSF R, His Tag (CSR-H5228) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Glu 512 (Accession # NP_005202.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.1 kDa. The protein migrates as 80-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 501657528 Supplier ACROBiosystems Inc Supplier No. CSRH5228200UG
Recombinant Protein;Human M-CSF R / CSF1R / CD115 Protein;200UG;Human M-CSF R, His Tag (CSR-H5228) is expressed from human 293 cells (HEK293). It contains AA Ile 20 - Glu 512 (Accession # NP_005202.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 55.1 kDa. The protein migrates as 80-95 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.