missing translation for 'onlineSavingsMsg'
Learn More

Sorbitan Monostearate, NF, 68-76%, Spectrum™ Chemical

Catalog No. 18606938
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
90 kg
Packaging:
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18606938 90 kg Poly Pail
18606936 2.5 kg Poly Pail
18606937 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18606938 Supplier Spectrum Chemical Mfg Cor Supplier No. SPA6390KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SPA63, 1338-41-6, C24H46O6

Spectrum™ Chemical Sorbitan Monostearate, NF is used as an emulsifier and nonionic surfactant in pharmaceutical formulations. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1338-41-6
Molecular Formula C24H46O6
Molecular Weight (g/mol) 430.63
MDL Number MFCD00005366,MFCD00005366
InChI Key HVUMOYIDDBPOLL-IIZJTUPISA-N
IUPAC Name 2-[(2R,3S,4R)-3,4-dihydroxyoxolan-2-yl]-2-hydroxyethyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(O)[C@H]1OC[C@@H](O)[C@@H]1O

Specifications

CAS Max % 1
Ignition Residue 0.005
Packaging Poly Pail
Quantity 90 kg
Saponification Value 147-157
Percent Purity 68 to 76%
Grade NF
Physical Form Powder or Waxy Solid
Chemical Name or Material Sorbitan Monostearate