Learn More
Enzo Life Sciences Mcl 1 polyclonal antibody (50 g)
Supplier: Enzo ADIAAP240D
Host: Rabbit. Immunogen: Synthetic peptide corresponding to a portion of human Mcl-1. UniProt: Q07820. Source: Purified from rabbit serum. Species Reactivity: Human, Rabbit, Porcine, Monkey, Guinea pig, Dog. Applications: IF, IHC (PS), WB. Recommended dilutions: Immunohistochemistry (paraffin sections, 1:50). Western Blot (1:500-1:1,000, ECL). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Peptide affinity purified. Formulation: Liquid. In PBS containing 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.