missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ McCoy 5A Medium, Iwakata and Grace Modification with L-Glutamine

Catalog No. mp91255354
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP91255354 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP91255354 Supplier MP Biomedicals™ Supplier No. 091255354
Only null left

Ideal for leukocytes, biopsy tissues, a broad range of human and rat normal and transformed cell types, and demanding primary and continuous cell lines

McCoy's media were originally formulated for the growth and support of human lymphocytes. The 5A modification originated as a reformulation of McCoy's by Hsu and Kellogg. It was then further modified by Iwakata and Grace featuring increased levels of i-inositol and glucose. This final modification produced a media identical to RPMI 1629. McCoy's 5A media supports the indefinite proliferation of Walker 256 carcinoma cells.

Specifications

Form Liquid
Product Type Cell Culture Media
Sterility Sterile
Without Additives No Glutamine
For Use With (Application) Mammalian Cell Culture
Quantity 500 mL
pH 6.6 to 7.2
Content And Storage Store at 4°C. Protect from light.
Description McCoy 5A Medium