missing translation for 'onlineSavingsMsg'
Learn More

MBTH Indicator, 0.05% (w/v), Ricca Chemical

APHA/ASTM for Cyanide Testing

Supplier:  Ricca Chemical Company 46224

Quantity 120 mL
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. 46224

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

Amber Glass Bottle
120 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents