Learn More
Sklar Instruments Mayo Dissecting Scissors, 6-3/4 in., Mayo Dissecting Scissors, Straight, Blunt/Blunt,EA
Catalog No. 501109737
Encompass
Mayo Dissecting Scissors, 6-3/4 in., Mayo Dissecting Scissors, Straight, Blunt/Blunt, Sklar Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved, in a variety of lengths, and have a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. Sklar also offers Mayo Dissecting Scissors with a selection of special features to meet procedure specifications and surgeon preferences. This product is straight with smooth, blunt/blunt tips, and an overall length of 6-3/4 inches.
Catalog No. 501109737 Supplier Sklar Instruments Supplier No. 151567
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Mayo Dissecting Scissors, 6-3/4 in., Mayo Dissecting Scissors, Straight, Blunt/Blunt, Sklar Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved, in a variety of lengths, and have a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. Sklar also offers Mayo Dissecting Scissors with a selection of special features to meet procedure specifications and surgeon preferences. This product is straight with smooth, blunt/blunt tips, and an overall length of 6-3/4 inches.