missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ MaxRecovery+™, Pipette Tip, Reload, Non-Sterile

Catalog No. 01677278
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
1000 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
01677278 200 μL
01677330 10 μL
01677199 1000 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01677278 Supplier Axygen™ Supplier No. PRFL200CL
Get Up to $2,500 in Products with Purchase
View all promotions

RNase-/DNase-free

  • For more than 20 years Axygen Universal pipette tips have set the standard for quality accuracy and precision in laboratories worldwide
  • Redesigned to meet every researcher needs the next generation of Axygen Universal Fit pipette tips deliver improved pipettor compatibility durable hinged racks and a complete offering from 10 μL to 1200 μL
  • Features include extended length tips wide bore designs for viscous solutions and proprietary MaxRecovery+™ technology to enhance liquid handling performance
  • RNaseDNase free
  • Nonpyrogenic
  • Human DNA free
  • ATP free
  • Available with MaxRecovery+ proprietary next generation low retention technology formulated with no intentionally added PFAS

Specifications

Volume (Metric) 200 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Autoclavable Autoclavable
Format Refill
Tip Style Beveled
Quantity 4800/Cs., 960/Pk.
Product Type Pipette Tips
Color Code Yellow
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated No
Material Polypropylene
Fit for Purpose Certified Quality
Show More Show Less