Learn More
Thermo Scientific™ Accessories for MaxQ™ 5000 Shakers
Increase the utility of the Thermo Scientific™ MaxQ™ 5000 Shaker with the MaxQ 5000 Gassing Manifold and Stainless-Steel Shelf.
Supplier:  Thermo Scientific™ SHK50006
 View more versions of this product
Catalog No. 11675460
Only null left
Explore available promotions
Description
Description
Increase the utility of the Thermo Scientific™ MaxQ™ 5000 Shaker with the MaxQ 5000 Gassing Manifold and Stainless-Steel Shelf.
Gassing Manifold
  • Enables direct gassing into flasks for anaerobic cultures
  • Offered with 8 or 16 outlets
  • Easy-access drain system offers convenience in draining and disinfecting
Stainless-Steel Shelf
  • For back wall of chamber
  • L × W: 33.25 × 4.0″ (84.5 × 10.2cm)
  • Retractable foot casters allow easy manipulation in lab
Specifications
Specifications
Shelf
Stainless steel shelf for chamber
1
84.5 x 10.2 cm
MaxQ 5000
Product Suggestions
Product Suggestions