missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ MaximaDry™ Diaphragm Vacuum Pump

Clean, reliable, oil-free vacuum pumps are ideal for low to high boiling point rotary evaporation and distillation, filtration, blotting, and solid phase extraction.

Supplier:  Fisherbrand™ 076857/029941

 Afficher plus de versions de ce produit

Numéro de catalogue. 1388020

Il en reste null

Exige : 0.375 in. I.D. tubing

Description

Description

Clean, reliable, oil-free vacuum pumps are ideal for high boiling point rotary evaporation and distillion, vacuum concentration, and larger vacuum ovens (>0.5 cu.ft.)

Quiet, reliable operation without the hazards of handling pump oil or contaminated water. Excellent replacement for rotary vane pumps and water aspirators, especially in applications where chemical vapors are encountered.

  • Withstands aggressive gases and vapors — can be used with aggressive solvents without the risk of damaging the pump
  • Single-piece PTFE coated diaphragm and molded PTFE pump heads provide chemical resistance
  • High-flow, multiport valves are made from tough FFPM fluoropolymer to withstand pumping heavy solvent vapors and liquid condensate without a loss in performance
  • Fan-cooled (TEFC) motor is rated for continuous operation and never needs lubrication
  • Compact design fits easily on a benchtop or in a fume hood
  • Setup and disassembly are quick and easy and do not require any special tools
  • Available in either 115 V/60 Hz or 230 V/50 Hz

Spécifications

Identifiants chimiques

IP44
15 psig

Spécifications

6 torr
16 x 7 x 9 in. (40.6 x 17.8 x 23 cm)
MaximaDry 35 to 6; 115V
60 Hz
IP44
15 psig
115 V 60 Hz
PTFE (Pump Head)
35 L/min.
Diaphragm Vacuum Pump
Vacuum Oven
115 V
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions