missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ MaximaDry™ Diaphragm Vacuum Pump

Catalog No. 1388018
Change view
Click to view available options
Flow Rate:
20 L/min.
35 L/min.
40 L/min.
Max. Vacuum:
1.5 torr
6 torr
65 torr
75 torr
Voltage:
115 V
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Flow Rate Max. Vacuum Voltage
1388018 20 L/min. 6 torr 115 V
1388014 20 L/min. 75 torr 115 V
1388022 35 L/min. 1.5 torr 115 V
1388020 35 L/min. 6 torr 115 V
1388016 40 L/min. 65 torr 115 V
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 1388018 Supplier Fisherbrand™ Supplier No. 076752/029940

Requires

0.375 in. I.D. tubing

Clean, reliable, oil-free vacuum pumps are ideal for low to high boiling point rotary evaporation and distillation, filtration, blotting, and solid phase extraction.

Clean, reliable, oil-free vacuum pumps are ideal for high boiling point rotary evaporation and distillion, vacuum concentration, and larger vacuum ovens (>0.5 cu.ft.)

Quiet, reliable operation without the hazards of handling pump oil or contaminated water. Excellent replacement for rotary vane pumps and water aspirators, especially in applications where chemical vapors are encountered.

  • Withstands aggressive gases and vapors — can be used with aggressive solvents without the risk of damaging the pump
  • Single-piece PTFE coated diaphragm and molded PTFE pump heads provide chemical resistance
  • High-flow, multiport valves are made from tough FFPM fluoropolymer to withstand pumping heavy solvent vapors and liquid condensate without a loss in performance
  • Fan-cooled (TEFC) motor is rated for continuous operation and never needs lubrication
  • Compact design fits easily on a benchtop or in a fume hood
  • Setup and disassembly are quick and easy and do not require any special tools
  • Available in either 115 V/60 Hz or 230 V/50 Hz

Plugs into electrical outlet; no hardwiring required.

Specifications

Max. Vacuum 6 torr
Electrical Requirements 115 V 60 Hz
Dimensions (L x W x H) 15 x 6.5 x 8.5 in. (38 x 16.5 x 22 cm)
Model MaximaDry 20 to 6 1;15V
Flow Rate 20 L/min.
Frequency 60 Hz
Pump Type Vacuum Pump
Enclosure Rating IP44
Applications Low Boiling Point Rotary Evaporation and Distillation
Maximum Pressure 15 psig
Voltage 115 V
Show More Show Less