missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Maxima Rotary Vane Vacuum Pumps - Explosion Proof

Fisherbrand™ Maxima rotary vane vacuum pumps extend service intervals due to robust materials and design features.

 View more versions of this product

Catalog No. 01184213

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Includes: Exhaust Filter, Oil Fill Funnel, Hose Clamp

Description

Description

Features:

  • Coating on pump internal components protect pump from aggressive chemicals. Increases service interval.
  • Large oil reservoir dilutes aggressive chemicals. Increases service interval.
  • Cooling system allows pump to run 10°C cooler than standard rotary vane pumps. The lower temperature leads to reduced chemical activity within the pump, slows down rates of oil consumption, extending service intervals.
  • Anti-Suckback valve prevents oil from being sucked into application in the event of a power failure.  
  • Gas ballast minimizes condensation of vapors in pump
  • Exhaust Filter, Funnel, and Hose Clamp are included in all Fisherbrand Maxima pump packages. 115V versions include US 115V cord. 230V verions include Shucko, UK, and US 230V Cord.

Certifications

UL

Warranty

1 Year, Materials and Workmanship

Specifications

Chemical Identifiers

Explosion Proof Rotary Vane
115 V, 60 Hz, 1 Ph, XPRF
Vacuum Manifolds, Vacuum Oven, Mass Spectroscopy, Backing Turbo Pumps, Sputter Coater, Tube Furnace
Exhaust Filter, Oil Fill Funnel, Hose Clamp
Explosion Proof Motor
115 V

Specifications

1.5x10⁻³ torr
Exhaust Filter, Oil Fill Funnel, Hose Clamp
115 V, 60 Hz, 1 Ph, XPRF
Inlet connection: NW16 or 5/8 to 3/4 in. ID Tubing
55 dBA
5.6 CFM
1 L
5 x 10-4 torr
Explosion Proof Rotary Vane
1740 RPM
22.4 x 7.1 x 9.1 in.
M8C
59.6 lbs
Explosion Proof Motor
Vacuum Manifolds, Vacuum Oven, Mass Spectroscopy, Backing Turbo Pumps, Sputter Coater, Tube Furnace
115 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions