missing translation for 'onlineSavingsMsg'
Learn More

PIP™ MaxiDry™ Zero™ Double-Dipped MicroFoam Nitrile-Coated Gloves with Thermal Lining

Catalog No. p-7180106
Change view
Click to view available options
Size:
Small
Large
X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19046436 Small
19046438 Large
19046439 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19046436 Supplier PIP™ Supplier No. GP56451S
Only null left

THERMtech™ technology platform offers thermal resistance up to -10°C (14°F) inside the glove under high activity, with a coating designed to remain flexible for temperature up to -30°C (-22°F).

  • Seamless construction increases comfort and breathability
  • MicroFoam Nitrile coating is compatible with light oils to enhance grip and abrasion resistance
  • Knit Wrist helps prevent dirt and debris from entering the glove
  • Liquid repellence keeps moisture away from the skin to ensure hands remain warm
  • High degree of thermal insulation without sacrificing flexibility, dexterity, weight, and comfort
  • Gloves help to prevent the effects of wind chills for outdoor applications
  • Coating remains flexible even in extremely cold environments
  • Optimized grip delivered though the ultra soft coating to ensure maximum flex and contact with the items being handled
  • Insulation liner is bonded away from flex and movement points of the hand (i.e. knuckles) to ensure comfort
  • All ingredients used in manufacturing and construction are compliant with the REACH legislation
  • Gloves are laundered prior to final packaging to ensure that they are clean and safe for use
  • The Oeko-Tex™ Association has evaluated and certified this product as skin safe from the second it comes into contact with the skin
  • The Skin Health Alliance has awarded professional dermatological accreditation following review of the scientific dossier behind this product
  • Complies with FDA food handling requirements 21 CFR, Part 177
  • Silicone-free

Specifications

Certifications/Compliance FDA 21 CFR 177.2600
Coating Material Nitrile
Color Purple
Exterior Finish Fully Textured
Hand Specific
Material Nylon Lycra
Product Type Cold-resistant Gloves
Size Small
For Use With (Application) Outdoor Construction/Maintenance, Agriculture, Cold Food Storage, and Chilled Distribution
Glove Cuff Style Knit
Liner Material Thermal
Series 56-451
Yarn Gauge 15
Length (English) 9.8 in.
Length (Metric) 25 cm
Show More Show Less