missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Maxi Pipet Tips
GREENER_CHOICE

Catalog No. 02707467
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
1 to 5 mL
1000 to 5000 μL
250 to 2500 μL
500 to 2500 μL
5000 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
02707467 1 to 5 mL
211953 1000 to 5000 μL
02681409 500 to 2500 μL
02681410 5000 μL
02707131 250 to 2500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 missing translation for 'options'
Catalog No. 02707467

For use with large volume pipets

  • Designed for use when transferring large sample volume
  • Advanced molding technology ensure tips are straight
  • Alternative option to serological pipets
  • Nonsterile; certified by independent laboratory to be RNase, DNase, DNA, and Pyrogen free

Specifications

Volume (Metric) 1 to 5 mL
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Length (Metric) 12.9 cm
Format Bulk
Tip Style Universal, Macro
For Use With (Equipment) Fisherbrand Elite, Fisherbrand II, Thermo Scientific
Material Polypropylene