missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Maxi Pipet Tips
GREENER_CHOICE

Catalog No. 02681410
Change view
Click to view available options
Volume (Metric):
1 to 5 mL
1000 to 5000 μL
250 to 2500 μL
500 to 2500 μL
5000 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
02681410 5000 μL
211953 1000 to 5000 μL
02707467 1 to 5 mL
02681409 500 to 2500 μL
02707131 250 to 2500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 02681410
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with large volume pipets

  • Designed for use when transferring large sample volume
  • Advanced molding technology ensure tips are straight
  • Alternative option to serological pipets
  • Nonsterile; certified by independent laboratory to be RNase, DNase, DNA, and Pyrogen free

Specifications

Volume (Metric) 5000 μL
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Length (English) 4.75 in.
Length (Metric) 150 mm
Format Bulk
Tip Style Universal, Macro
For Use With (Equipment) Eppendorf Research, Jencons Sealpette, Genemate ISC Bio Express
Graduated No
Material Polypropylene
Show More Show Less