Learn More
BD MAX™ Molecular Swab Collection Kit
Catalog No. B443925
Change view
Click to view available options
Type:
Molecular swabs for collection
Catalog No. Type
B443925 Molecular swabs for collection
1 options
Catalog No. B443925 Supplier BD Supplier No. 443925
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This Molecular Swab Collection Kit is intended to be used in clinical settings for collection and transport of vaginal and endocervical swab specimens.
  • Molecular Swab Sample Buffer Tubes containing 2 mL Sample Buffer for collection and transport of vaginal and endocervical swab specimens.
  • BD Molecular Collection Swabs are for use with BD Molecular products
  • BD Molecular Collection Swab is a sterile polyester swab with a shaft that is scored for breaking
Specifications
Type Molecular swabs for collection