missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Matrix™ 1.0mL ScrewTop Tubes
GREENER_CHOICE

Catalog No. 14754579
Change view
Click to view available options
Closure Color:
Blue
Colorless
Green
Not Included
Purple
Red
White
Yellow
Color:
Amber
Clear
Packaging:
5 Latch Racks of 96 Tubes
Bulk, 480 Tubes
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Color Color Packaging
14754579 Not Included Amber 5 Latch Racks of 96 Tubes
14754550 Colorless Clear Bulk, 480 Tubes
14754554 Red Clear Bulk, 480 Tubes
14754557 Yellow Clear Bulk, 480 Tubes
14754551 Blue Clear Bulk, 480 Tubes
14754552 Green Clear Bulk, 480 Tubes
14754555 White Clear Bulk, 480 Tubes
14754553 Purple Clear Bulk, 480 Tubes
14754558 Colorless Clear 5 Latch Racks of 96 Tubes
14754568 Red Clear 5 Latch Racks of 96 Tubes
14754576 Yellow Clear 5 Latch Racks of 96 Tubes
14754561 Blue Clear 5 Latch Racks of 96 Tubes
14754564 Green Clear 5 Latch Racks of 96 Tubes
14754570 White Clear 5 Latch Racks of 96 Tubes
14754566 Purple Clear 5 Latch Racks of 96 Tubes
14754578 Not Included Clear 5 Latch Racks of 96 Tubes
14754559 Red Amber 5 Latch Racks of 96 Tubes
14754572 Colorless Clear 5 Latch Racks of 96 Tubes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 14754579 Supplier Thermo Scientific™ Supplier No. 3742AMB
Only null left

Store samples securely at low temperatures, including vapor phase liquid nitrogen, with these 1.0mL screw-top tubes that include color-coded caps to simplify storage.

Securely store samples at low temperatures, including vapor phase Liquid Nitrogen with Thermo Scientific™ Matrix™ 1.0mL ScrewTop Tubes. These tubes are molded from medical grade resins and feature a permanent, high contrast 2D barcode. Used in combination with Thermo Scientific™ VisionMate™ 2D barcode readers, manual and automated capping equipment, and labeling options, the 2D ScrewTop Tubes meet a range of manual and automated long term storage needs.

Features:

Dependable Sample Protection

  • Tubes are constructed of virgin class VI medical grade polypropylene
  • Screw-top caps feature a soft silicone gasket that conforms to mating surfaces to ensure sample preservation
  • Racks are constructed of sturdy acrylonitrile butadiene styrene (ABS) to protect tubes in case of accidental dropping
  • Screw-top tubes are compatible with storage down to -196°C

Secure Tracking

  • Permanent, unique 2D barcode is laser-etched on the bottom of each tube
  • 2D barcode allows you to scan and decode tubes at once without removing them from the rack, and allows data to be associated with each tube
  • Compatible with our range of VisionMate readers

Superior Storage Format

  • ANSI-format latch racks maximize storage space and are compatible with multichannel manual pipettes or automated liquid handling equipment
  • Several capping/decapping options available to manipulate a single tube, 8 tubes, or 96 tubes at a time

Stringent Quality Control

  • Every 2D barcode is scanned for readability
  • Every 2D barcode is checked against the database of all previous Thermo Scientific 2D barcodes to ensure there are zero duplicates
  • Each tube is leak tested to ensure tube integrity and sample security
  • All tubes are supplied free from DNA, RNase, DNase, endotoxins and cytotoxins

Specifications

Color Amber
Closure Color Not Included
Capacity (Metric) 1.0 mL
Material Polypropylene
Packaging 5 Latch Racks of 96 Tubes
Product Line Matrix
Sterility Sterile
Closure Included No
Capacity (English) 0.0338 fl oz.
Dnase Free Yes
Closure Type Screw Cap
Fit for Purpose Sterile and PCR Ready
Product Type Screw Top Tube
Show More Show Less
Do you offer Thermo Scientific storage tubes for storing light-sensitive samples?

We offer the following 0.5 mL and 1.0 mL Matrix 2D ScrewTop amber-colored resin storage tubes with red screw caps:
Cat. No.; Description; Quantity 3741AMB; 1.0 mL, Amber, 2D Barcoded, V-bottom, ScrewTop with caps, Sterile; 5 latch racks of 96/case 3742AMB; 1.0 mL, Amber, 2D Barcoded, V-bottom, ScrewTop without caps, Sterile; 5 latch racks of 96/case 3743AMB; 0.5 mL, Amber, 2D Barcoded, V-bottom, ScrewTop with caps, Sterile; 5 latch racks of 96/case 3745AMB; 0.5 mL, Amber, 2D Barcoded, V-bottom, ScrewTop without caps, Sterile; 5 latch racks of 96/case