missing translation for 'onlineSavingsMsg'
Learn More

GVS™ Wallerstein Broth (WL) 2mL Ampoules (50 Pack)

Catalog No. 10486143
Change view
Click to view available options
Broth Type:
Wallerstein Differential Broth
Wallerstein Nutrient Broth
Detects:
Bacterial counts
Yeast growth and counts
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Broth Type Detects
10486143 Wallerstein Nutrient Broth Yeast growth and counts
10486144 Wallerstein Differential Broth Bacterial counts
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10486143 Supplier GVS™ Supplier No. 10496108

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready-to-use 2mL ampoule Wallerstein Broth (WL) is a liquid culture growth media that can save time and money by shortening prep time and minimizing possible contamination. The 2mL ampoules contain enough media to saturate a standard absorbent pad in a petri dish

Description

  • WL Nutrient Broth is for the cultivation and enumeration of yeast and WL Differential Broth is for determination of bacterial count. Use of the medium at pH 5.5 and incubation at 25°C will give reliable counts for brewer’s yeast. Adjustment of the pH to 6.5 and incubation at 30°C allows for the selective growth of baker’s and distiller’s yeast

Features and Benefits

  • Pre-Sterilized and Ready to Use
  • Time Saving, 2 mL Portions, Reduce Risk of Cross-Contamination.
  • 2mL ampoules contain enough media to saturate a standard absorbent pad in a petri dish
  • Optimal media stability, sterility, and reproducibility
  • Less time-consuming, higher productivity
  • Batch-specific quality certificate in each pack, Confirms Reproducibility and Reliability

Applications

  • Microbiological analysis.
  • Use with membrane filters to culture bacteria, yeasts, and molds.

Liquid Media
Ready-to-use media considerably reduce the preparation time in quality control laboratories and also effectively reduce the risks of cross contamination. GVS is cooperating closely with quality assurance managers in the industry in the development of its own media and test kits. This intensive product development has produced a range of products that is being used to monitor production plants and conduct Microbiological checks on raw materials through to final product release in laboratories in more than 40 countries.

See 'GVS Life Sciences Microbiology Testing mpg YouTube' Video for further details on use.

Specifications

Broth Type Wallerstein Nutrient Broth
Certifications/Compliance Batch-specific quality certificate in each pack
Detects Yeast growth and counts
Manufacturing Origin United States
Sterility Sterile
Incubation Conditions pH 5.5 at 25°C for brewer's yeast. pH 6.5 at 30°C for baker's and distiller's yeast
No. per Pack 50
Packaging Type Ampoules
Quantity 2 mL