missing translation for 'onlineSavingsMsg'
Learn More

Matrix Modifier, 5% (w/v) Ammonium Nitrate in 2% (v/v) HNO3, Ricca Chemical

Catalog No. MMAN5N50
Change view
Click to view available options
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
MMAN5N50 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MMAN5N50 Supplier Ricca Chemical Company Supplier No. MMAN5N50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Matrix Modifier
Packaging Natural Poly Bottle
Name Note Ammonium Nitrate
CAS Min % 91.0, 4.54, 2.73
Concentration or Composition Notes 5%
CAS Max % 94.0, 4.72, 2.84
CAS 6484-52-2, 7697-37-2