missing translation for 'onlineSavingsMsg'
Learn More

Matrix Modifier, 2.0% Mg (20,000 ppm Mg), Ricca Chemical

Catalog No. MMMG20KN50
Change view
Click to view available options
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
MMMG20KN50 Natural Poly Bottle
MMMG20KN100 Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MMMG20KN50 Supplier Ricca Chemical Company Supplier No. MMMG20KN50

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Matrix Modifier
Packaging Natural Poly Bottle
Concentration or Composition Notes 2.0% Pd,20,000ppm Pd
CAS Min % 76.0, 16.09, 5.72
CAS Max % 79.0, 16.74, 5.96
CAS 13446-18-9, 7697-37-2