missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Matrix Modifier, 0.5% Pd (5,000 ppm Pd), Ricca Chemical
Specifications
Specifications
| Chemical Name or Material | Matrix Modifier |
| Packaging | Natural Poly Bottle |
| Concentration or Composition Notes | 0.5% Pd,5000ppm Pd |
| CAS Min % | 88.0, 9.29, 0.35, 0.06 |
| CAS Max % | 92.0, 9.67, 0.36, 0.06 |
| CAS | 7697-37-2, 7440-05-3, 7647-01-0 |