missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Matrigel™ Matrix for Organoid Culture
GREENER_CHOICE

Catalog No. 08774406
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08774406 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08774406 Supplier Corning™ Supplier No. 356255
  • ONLINE EXCLUSIVE PRICE
    $1,188.44
    $687.77 / Each

    Save $500.67

    42.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Corning™ Matrigel™ Matrix for Organoid Culture is a solubilized basement membrane preparation that has been optimized for organoid culture.

Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

Corning Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Specifications

Concentration Lot dependent
Product Type Stem Cell Media
For Use With (Application) Organoid Culture and workflow
Quantity 10 mL
Source Engelbreth-Holm-Swarm (EHS) mouse tumor
Content And Storage -20°C