missing translation for 'onlineSavingsMsg'
Learn More

Corning™ Matrigel™ Matrix-3D Plate, 384-well
GREENER_CHOICE

Catalog No. 08774410
Change view
Click to view available options
Color:
Black
White, Clear
No. per Pack:
1
5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color No. per Pack
08774410 White, Clear 1
08774408 Black 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 08774410 Supplier Corning™ Supplier No. 356258

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Corning™ Matrigel™ Matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma. It has a high-throughput format that can be used for drug discovery with 3D cell cultures such as organoids, spheroids or tumoroids.

  • A high-throughput pre-coated format that contains Matrigel™ and can be used for drug discovery applications with 3D cell cultures such as organoids, spheroids or tumoroids
  • Extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparin sulfate proteoglycans, entactin/nidogen, and a number of growth factors
  • Each lot is verified for intraplate consistency
  • Tested and found negative for the presence of bacteria and fungi
  • Easy and convenient as Matrigel™ is pre-dispensed into each well
  • Formats provide intra-plate consistency
  • Allows for sandwich/overlay and embedded 3D application workflows

Specifications

Barcode No Barcode
Breakapart No
Lid With Lid
Content And Storage Store at -20°C
Autoclavable Not Autoclavable
Color White, Clear
Disposable Single-use
Material Polystyrene
No. of Wells 384
Surface Treatment Matrigel Matrix
Well Shape Flat Bottom (F-well)
Dnase Free No
For Use With (Application) 3D cell culture
Includes Guidelines for Use
No. per Case 1
No. per Pack 1
Regulatory Status RUO
Rimmed No
RNase Free No
Skirt Color White with clear bottom
Show More Show Less