missing translation for 'onlineSavingsMsg'
Learn More

Corning Matrigel™ Growth Factor Reduced (GFR) Basement Membrane Matrix, Phenol Red-free, LDEV-free

Catalog No. C6000025301
Change view
Click to view available options
Concentration:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
C6000025301 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C6000025301 Supplier Corning Supplier No. 6000025301

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.

  • Matrigel matrix is a solubilized basement membrane preparation extracted from the Engelbreth-Holm-Swarm (EHS) mouse sarcoma, a tumor rich in extracellular matrix proteins, including Laminin (a major component), Collagen IV, heparan sulfate proteoglycans, entactin/nidogen, and a number of growth factors.

Specifications

Cell Type 2D and 3D
Classification Animal Origin, Reduced Growth Factor
Concentration 5 mL
Form Frozen
Product Type Extracellular Matrix
Serum Level Serum-free
For Use With (Application) Cell Growth and Differentiation, Metabolism and Toxicology Studies, Invasion Assays, in vitro and in vivo Angiogenesis Assays, in vivo Angiogenesis Studies and Augmentation of Tumors in Immunosuppressed Mice
Manufacturing Quality 9001
Product Line Corning Matrigel
Quantity 5 mL
Shipping Condition -20°C
Assays Angiogenesis, Invasion Assays and drug screening in 3D
Certifications/Compliance Research Use Only
Endotoxin Level >16 EU/mL
Format Frozen
Formulation Dulbecco's Modified Eagle's Medium with 50 μg/mL gentamycin.
Included Antibiotics Gentamycin
Regulatory Status RUO
Source Engelbreth-Holm-Swarm (EHS) Mouse Sarcoma
Species Mouse
Stem Cell Qualified No
Tested For Bacteria, Fungi, Mycoplasma
Color Clear
Content And Storage -20°C
Culture Type 3D Cell Culture, Feeder-free Stem Cell Culture (Human, iPS - Induced Pluripotent Stem, Embryonic), Adherent Cell Culture, Mammalian Cell Culture
Show More Show Less