Learn More
Eisco™ Material Solids Block Kit
The Material Solids Block Kit is intended to familiarize students with appearance, feel, texture and density of these common materials.
Supplier:  Eisco™ PH0114A
Catalog No. S12928
Only null left
Description
Description
  • 14 Different Blocks
  • Kit contains 11 different materials
  • This kit is intended to familiarize students with appearance, feel, texture and density of these common materials
  • All measurements are approx. and may variable from kit to kit
  • Aluminum block 2 in. x 1.5 in. 1.125 in.
  • Measurements: Aluminum Rectangle 3.875 in. x 0.75 in. x 0.75 in.; Brass Block 2 in. x 0.75 in. x 0.75 in.; Styrofoam block 2 in. x 1.5 in. 1.125 in.; Wax Block 2 in. x 1.5 in. 1.125 in.; Stainless Steel Block 2 in. x 2 in. x 0.75; Wood Block 2 in. x 1.5625 in. x 1.1875; Wood Rectangle; 3.875 in. x 0.75 in. x 0.75 in.; Marble Rectangle 3.875 in. x 0.75 in. x 0.75 in.; Glass Rectangle 3.875 in. x 0.75 in. x 0.75 in.; Acrylic Rectangle 3.875 in. x 0.75 in. x 0.75 in.; Steel Block 1.5 in. x 1.5 in. 0.75 in.; Steel Block 2 in. x 1.5 in. 1.125 in.
Specifications
Specifications
14 Pk.
Density Sample Set
Product Suggestions
Product Suggestions