missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Wanzl Material Handling Drop Down Dolly
Description
- Improved Efficiency: Six (6) or Eight (8) Tote/Bin capacity, Storage Bin for miscellaneous items and Two (2) storage hooks for handing scanners or bags
- Versatile Tote Placement: Adjustable-height brackets allow use of totes with different heights (1 in. incremental height adjustments )
- Tote Security: Totes are held in place with raised ledges on the adjustable height brackets to prevent the totes from sliding off
- Top Shelf Storage: Top Wire Shelf has a raised ledge to keep products secured and preventing products from sliding off
- Ergonomic: Dual ergonomic handles to safely maneuver the cart
- Corner Bumpers: eliminates cart and store fixture damage
- Durable: Heavy-duty steel construction
Specifications
Specifications
| Color | Chrome |
| For Use With (Equipment) | Multi-Pick Cart |
| Material | Steel (18 ga.) |